missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
3-Methylthiopropylamine 98.0+%, TCI America™
Supplier: TCI America M075425ML
Specifications
3-Methylthiopropylamine | |
Yellow | |
C4H11NS | |
25 mL | |
3-methylthiopropylamine, 3-methylthio propylamine, 1-propanamine, 3-methylthio, 3-methylthiopropanamine, methylmercaptopropylamine, unii-1iap0cgd5s, 3-methylmercaptopropylamine, 3-methylthio-1-propanamine, 3-methylsulfanyl propan-1-amine, 1iap0cgd5s | |
CSCCCN | |
105.199 | |
CHEBI:16696 | |
≥98.0% (GC,T) |
4104-45-4 | |
170°C | |
MFCD00041898 | |
2735 | |
KKYSBGWCYXYOHA-UHFFFAOYSA-N | |
3-methylsulfanylpropan-1-amine | |
77743 | |
105.20 | |
Liquid |
Chemical Identifiers
4104-45-4 | |
105.199 | |
KKYSBGWCYXYOHA-UHFFFAOYSA-N | |
77743 | |
3-methylsulfanylpropan-1-amine |
C4H11NS | |
MFCD00041898 | |
3-methylthiopropylamine, 3-methylthio propylamine, 1-propanamine, 3-methylthio, 3-methylthiopropanamine, methylmercaptopropylamine, unii-1iap0cgd5s, 3-methylmercaptopropylamine, 3-methylthio-1-propanamine, 3-methylsulfanyl propan-1-amine, 1iap0cgd5s | |
CHEBI:16696 | |
CSCCCN |
Safety and Handling
TSCA : Yes