missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Methylthiophene-2-carboxamide, 99%

$57.36 - $200.47

Chemical Identifiers

CAS 76655-99-7
Molecular Formula C6H7NOS
Molecular Weight (g/mol) 141.188
MDL Number MFCD01208055
InChI Key QZDTWJRYMXQXBX-UHFFFAOYSA-N
Synonym 3-methyl-2-thiophenecarboxamide, pubchem16135, 2-thiophenecarboxamide,3-methyl, 3-methyl-thiophene-2-carboxamide, 3-methyl-2-thiophenecarboxamide #, 2-thiophenecarboxamide, 3-methyl-9ci
PubChem CID 588128
IUPAC Name 3-methylthiophene-2-carboxamide
SMILES CC1=C(SC=C1)C(=O)N
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL1809206 View Documents Alfa Aesar™
L1809206
5g Each for $200.47
 
AAL1809203 View Documents Alfa Aesar™
L1809203
1g Each for $57.36
 
Specifications

Chemical Identifiers

76655-99-7
141.188
QZDTWJRYMXQXBX-UHFFFAOYSA-N
588128
CC1=C(SC=C1)C(=O)N
C6H7NOS
MFCD01208055
3-methyl-2-thiophenecarboxamide, pubchem16135, 2-thiophenecarboxamide,3-methyl, 3-methyl-thiophene-2-carboxamide, 3-methyl-2-thiophenecarboxamide #, 2-thiophenecarboxamide, 3-methyl-9ci
3-methylthiophene-2-carboxamide

Specifications

122°C to 125°C
76655-99-7
MFCD01208055
3-methyl-2-thiophenecarboxamide, pubchem16135, 2-thiophenecarboxamide,3-methyl, 3-methyl-thiophene-2-carboxamide, 3-methyl-2-thiophenecarboxamide #, 2-thiophenecarboxamide, 3-methyl-9ci
CC1=C(SC=C1)C(=O)N
141.188
141.19
3-Methylthiophene-2-carboxamide
5g
C6H7NOS
116188
QZDTWJRYMXQXBX-UHFFFAOYSA-N
3-methylthiophene-2-carboxamide
588128
99%
Safety and Handling

Safety and Handling

GHS H Statement
H301
Toxic if swallowed.

GHS P Statement
P264-P270-P301+P310-P321-P405-P501a
Wash thoroughly after handling.
Do not eat, drink or smoke when using this product.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Specific treatment (see label).
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger