missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Methylthiophene-2-boronic acid, 98%

Manufacturer:  Alfa Aesar™ H5320103

 View more versions of this product

Catalog No. AAH5320103



Specifications

Specifications

3-Methylthiophene-2-boronic acid
1g
C5H7BO2S
3-methylthiophene-2-boronic acid, 3-methylthiophen-2-yl boronic acid, 3-methyl-2-thienylboronic acid, 3-methylthiophen-2-yl-2-boronic acid, 3-methyl-2-thiopheneboronic acid, 3-methyl-2-thienyl boronic acid, 3-methyl-thiophen-2-boronic acid, 3-methylthiophene-2-boronicacid, acmc-209ede, boronic acid, 3-methyl-2-thienyl
B(C1=C(C=CS1)C)(O)O
141.979
142.01
192°C to 196°C
177735-09-0
MFCD00236049
MOOOGTOEKZWEJA-UHFFFAOYSA-N
(3-methylthiophen-2-yl)boronic acid
3276437
98%

Chemical Identifiers

177735-09-0
141.979
MOOOGTOEKZWEJA-UHFFFAOYSA-N
3276437
B(C1=C(C=CS1)C)(O)O
C5H7BO2S
MFCD00236049
3-methylthiophene-2-boronic acid, 3-methylthiophen-2-yl boronic acid, 3-methyl-2-thienylboronic acid, 3-methylthiophen-2-yl-2-boronic acid, 3-methyl-2-thiopheneboronic acid, 3-methyl-2-thienyl boronic acid, 3-methyl-thiophen-2-boronic acid, 3-methylthiophene-2-boronicacid, acmc-209ede, boronic acid, 3-methyl-2-thienyl
(3-methylthiophen-2-yl)boronic acid
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

SDS Documents