missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-(Methylthio)propylamine, 97%, ACROS Organics™

Chemical Identifiers

CAS 4104-45-4
Molecular Formula C4H11NS
Molecular Weight (g/mol) 105.2
MDL Number MFCD00041898
InChI Key KKYSBGWCYXYOHA-UHFFFAOYSA-N
Synonym 1-propanamine, 3-methylthio, 1iap0cgd5s, 3-methylmercaptopropylamine, 3-methylsulfanyl propan-1-amine, 3-methylthio propylamine, 3-methylthio-1-propanamine, 3-methylthiopropanamine, 3-methylthiopropylamine, methylmercaptopropylamine, unii-1iap0cgd5s
PubChem CID 77743
ChEBI CHEBI:16696
IUPAC Name 3-methylsulfanylpropan-1-amine
SMILES CSCCCN
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC414930010
SDS View Documents
Thermo Fisher Scientific
AC414930010
1g Glass bottle N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications

Chemical Identifiers

4104-45-4
105.2
KKYSBGWCYXYOHA-UHFFFAOYSA-N
77743
3-methylsulfanylpropan-1-amine
C4H11NS
MFCD00041898
1-propanamine, 3-methylthio, 1iap0cgd5s, 3-methylmercaptopropylamine, 3-methylsulfanyl propan-1-amine, 3-methylthio propylamine, 3-methylthio-1-propanamine, 3-methylthiopropanamine, 3-methylthiopropylamine, methylmercaptopropylamine, unii-1iap0cgd5s
CHEBI:16696
CSCCCN

Specifications

0.9600g/mL
Authentic
1.4920 to 1.4940
169.0°C
1g
96.0
97%
CH3SCH2CH2CH2NH2
04, IV, 1655
KKYSBGWCYXYOHA-UHFFFAOYSA-N
3-methylsulfanylpropan-1-amine
77743
105.2
97%
62°C
Glass bottle
0.96
Colorless
4104-45-4
100.0
C4H11NS
MFCD00041898
1-propanamine, 3-methylthio, 1iap0cgd5s, 3-methylmercaptopropylamine, 3-methylsulfanyl propan-1-amine, 3-methylthio propylamine, 3-methylthio-1-propanamine, 3-methylthiopropanamine, 3-methylthiopropylamine, methylmercaptopropylamine, unii-1iap0cgd5s
CSCCCN
105.2
CHEBI:16696
Liquid
3-(Methylthio)propylamine
Safety and Handling

Safety and Handling

GHS H Statement
Causes serious eye damage.
Causes severe skin burns and eye damage.

GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsi

GHS Signal Word: Danger

EINECSNumber : 223-875-2