Please login to your online account to display your discounted pricing

3-(Methylthio)propylamine, 97%, ACROS Organics™


Quantity 5g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  414930050

 View more versions of this product

Catalog No. AC414930050

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥96.0%
Physical Form Liquid
Formula Weight 105.2
Quantity 5g
Color Clear colorless
Beilstein 04, IV, 1655
Density 0.9600g/mL
CAS Min % 96.0
CAS Max % 100.0
Chemical Name or Material 3-(Methylthio)propylamine
Infrared Spectrum Authentic
Refractive Index 1.4920 to 1.4940 (20°C, 589nm)
Assay Percent Range 97%
Molecular Formula C4H11NS
Linear Formula CH3SCH2CH2CH2NH2
CAS 4104-45-4
Health Hazard 1 GHS Signal Word: Danger
Health Hazard 2 GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.
Health Hazard 3 GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Packaging Glass bottle
MDL Number MFCD00041898

Specifications

Chemical Name or Material 3-(Methylthio)propylamine
CAS 4104-45-4
CAS Min % 96.0
CAS Max % 100.0
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C4H11NS
Linear Formula CH3SCH2CH2CH2NH2
Formula Weight 105.2
MDL Number MFCD00041898
Accessories

GHS Signal Word: Danger

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.

SDS & Safety Documents