missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-(Methylthio)propionyl Chloride, TCI America™

$157.36 - $1,365.86

Chemical Identifiers

CAS 7031-23-4
Molecular Formula C4H7ClOS
Molecular Weight (g/mol) 138.609
MDL Number MFCD00059482
InChI Key NRHMXMBVLXSSAX-UHFFFAOYSA-N
Synonym 3-(Methylmercapto)propionyl Chloride
PubChem CID 12921440
IUPAC Name 3-methylsulfanylpropanoyl chloride
SMILES CSCCC(=O)Cl
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
M080625ML View Documents TCI America
M080625ML
25mL Each for $157.36
 
M0806500ML View Documents TCI America
M0806500ML
500mL Each for $1,365.86
 
Specifications

Chemical Identifiers

7031-23-4
138.609
NRHMXMBVLXSSAX-UHFFFAOYSA-N
12921440
CSCCC(=O)Cl
C4H7ClOS
MFCD00059482
3-(Methylmercapto)propionyl Chloride
3-methylsulfanylpropanoyl chloride

Specifications

96°C
25mL
C4H7ClOS
3265
NRHMXMBVLXSSAX-UHFFFAOYSA-N
3-methylsulfanylpropanoyl chloride
12921440
Clear Liquid at 20°C
3-(Methylthio)propionyl Chloride
Yellow
7031-23-4
MFCD00059482
3-(Methylmercapto)propionyl Chloride
CSCCC(=O)Cl
138.609
138.61
≥98.0% (T)
Safety and Handling

Safety and Handling

TSCA : No