missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
3-(Methylthio)propionyl Chloride 98.0+%, TCI America™
Supplier: TCI America M0806500ML
Specifications
3-(Methylthio)propionyl Chloride | |
Yellow | |
C4H7ClOS | |
500 mL | |
3-(Methylmercapto)propionyl Chloride | |
CSCCC(=O)Cl | |
138.609 | |
138.61 | |
Liquid |
7031-23-4 | |
96°C | |
MFCD00059482 | |
3265 | |
NRHMXMBVLXSSAX-UHFFFAOYSA-N | |
3-methylsulfanylpropanoyl chloride | |
12921440 | |
≥98.0% (T) |
Chemical Identifiers
7031-23-4 | |
138.609 | |
NRHMXMBVLXSSAX-UHFFFAOYSA-N | |
12921440 | |
CSCCC(=O)Cl |
C4H7ClOS | |
MFCD00059482 | |
3-(Methylmercapto)propionyl Chloride | |
3-methylsulfanylpropanoyl chloride |
Safety and Handling
TSCA : No