missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-(Methylthio)propionic Acid, TCI America™

Manufacturer:  TCI America M0811500G

 View more versions of this product

Catalog No. M0811500G



Specifications

Specifications

3-(Methylthio)propionic Acid
Yellow
646-01-5
MFCD00059635
3-methylthio propionic acid, 3-methylthiopropionate, 3-methylthiopropionic acid, 4-thiapentanoic acid, 3-methylsulfanyl propanoic acid, propanoic acid, 3-methylthio, 3-methylmercaptopropionic acid, unii-f523ali47d, 3-methylthio propanoic acid, propionic acid, 3-methylthio
CSCCC(=O)O
120.166
CHEBI:1438
Clear Liquid at 20°C
130°C
500g
C4H8O2S
3265
CAOMCZAIALVUPA-UHFFFAOYSA-N
3-methylsulfanylpropanoic acid
563
120.17
≥98.0% (GC,T)

Chemical Identifiers

646-01-5
120.166
CAOMCZAIALVUPA-UHFFFAOYSA-N
563
3-methylsulfanylpropanoic acid
C4H8O2S
MFCD00059635
3-methylthio propionic acid, 3-methylthiopropionate, 3-methylthiopropionic acid, 4-thiapentanoic acid, 3-methylsulfanyl propanoic acid, propanoic acid, 3-methylthio, 3-methylmercaptopropionic acid, unii-f523ali47d, 3-methylthio propanoic acid, propionic acid, 3-methylthio
CHEBI:1438
CSCCC(=O)O
Safety and Handling

Safety and Handling

TSCA : No

Documents