missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-(Methylthio)propionaldehyde, 98%

Manufacturer:  Alfa Aesar™ A1477430

 View more versions of this product

Catalog No. AAA1477430



Description

Description

Applications
3-(Methylthio)propionaldehyde, is used as food flavour ,for making Baked food, spices, soft drinks, candy. It is also used as an intermediate. It has wide applications in organic synthesis as solvents as well as reagents. They are used in the synthesis of specific compound classes include agricultural chemical, property-enhancing additives, pharmacological drugs, chemical resistant polymers, detergents, and rubber antioxidants. Some examples of aliphatic and aromatic thioethers found in flavouring agents

Solubility
It is readily soluble in alcohol solvents such as ethanol, propylene and glycol oil. It is insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Recommended storage temperature 2 - 8°C. Air sensitive. Handle and store under inert gas. Keep away from strong oxidizing agents, strong bases.
Specifications

Specifications

3-(Methylthio)propionaldehyde
Strong
-58°C
3268-49-3
CH3S(CH2)2CHO
UN2785
methional, 3-methylthio propionaldehyde, 3-methylthio propanal, propanal, 3-methylthio, 4-thiapentanal, 3-methylsulfanyl propanal, 3-methylmercapto propionaldehyde, 3-methylthiopropanal, propionaldehyde, 3-methylthio, 3-methylmercaptopropyl aldehyde
CSCCC=O
104.167
CHEBI:49017
98%
64°C (147°F)
165°C to 166°C
250g
C4H8OS
MFCD00007022
1739289
CLUWOWRTHNNBBU-UHFFFAOYSA-N
3-methylsulfanylpropanal
18635
104.17

Chemical Identifiers

3268-49-3
104.167
CLUWOWRTHNNBBU-UHFFFAOYSA-N
18635
3-methylsulfanylpropanal
C4H8OS
MFCD00007022
methional, 3-methylthio propionaldehyde, 3-methylthio propanal, propanal, 3-methylthio, 4-thiapentanal, 3-methylsulfanyl propanal, 3-methylmercapto propionaldehyde, 3-methylthiopropanal, propionaldehyde, 3-methylthio, 3-methylmercaptopropyl aldehyde
CHEBI:49017
CSCCC=O
Safety and Handling

Safety and Handling

GHS H Statement
H302-H312-H332-H315-H319-H317-H335-H227
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.
May cause respiratory irritation.
Combustible liquid.

GHS P Statement
P280h-P305+P351+P338-P309-P310
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

Warning

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: 4-THIAPENTANAL

RTECSNumber : UE2285000

TSCA : TSCA

Recommended Storage : 2° to 8°C

SDS Documents