Please login to your online account to display your discounted pricing

3-(Methylthio)propionaldehyde 95%, ACROS Organics™


Quantity 25g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC414920250

 View more versions of this product

Catalog No. AC414920250

  • / Each
 

Description & Specifications

Chemical Identifiers

Assay ≥96%
Formula Weight 104.17
Physical Form Liquid
Quantity 25g
Color Clear colorless
RTECS Number UE2285000
TSCA TSCA
EINECS Number 221-882-5
Density 1.05g/mL
CAS Min % 96.0
CAS Max % 100.0
Infrared Spectrum Authentic
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Packaging Glass bottle
Health Hazard 2 GHS H Statement:
Harmful in contact with skin.
Harmful if swallowed.
Causes severe skin burns and eye damage.
Health Hazard 1 GHS Signal Word: Danger
Assay Percent Range 97%
CAS 3268-49-3
Chemical Name or Material 3-(Methylthio)propionaldehyde
Linear Formula CH3SCH2CH2CHO
Name Note 95%
Molecular Formula C4H8OS

Specifications

Chemical Name or Material 3-(Methylthio)propionaldehyde
Name Note 95%
CAS 3268-49-3
CAS Min % 96.0
CAS Max % 100.0
Assay ≥96%
Assay Percent Range 97%
Molecular Formula C4H8OS
Linear Formula CH3SCH2CH2CHO
Formula Weight 104.17
Accessories

GHS Signal Word: Danger

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful in contact with skin.
Harmful if swallowed.
Causes severe skin burns and eye damage.

SDS & Safety Documents