Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-(Methylthio)phenylacetic acid, 95%

$80.50 - $229.35

Chemical Identifiers

Chemical Name or Material 3-(Methylthio)phenylacetic acid
CAS 18698-73-2
Molecular Formula C9H10O2S
Formula Weight 182.24
MDL Number MFCD03701554
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2745203

 
alfa aesar™
H2745203
1g Each for $80.50

AAH2745206

 
alfa aesar™
H2745206
5g Each for $229.35
Description & Specifications

Specifications

Formula Weight 182.24
Chemical Name or Material 3-(Methylthio)phenylacetic acid
Assay 95%
Molecular Formula C9H10O2S
MDL Number MFCD03701554
CAS 18698-73-2
Health Hazard 1 Warning
Melting Point 77° to 81°C
Quantity 1g
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.

Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.