missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-(Methylthio)phenyl isothiocyanate, 97%

Manufacturer:  Alfa Aesar™ L1299906

 View more versions of this product

Catalog No. AAL1299906



Specifications

Specifications

3-(Methylthio)phenyl isothiocyanate
144°C to 145°C (8mmHg)
51333-80-3
MFCD00041079
2208144
IHKAGFVCXPFGRP-UHFFFAOYSA-N
1-isothiocyanato-3-methylsulfanylbenzene
142841
97%
>110°C (230°F)
5g
C8H7NS2
UN2922
3-methylthio phenyl isothiocyanate, 1-isothiocyanato-3-methylsulfanyl benzene, 3-isothiocyanatophenyl methyl sulfane, 1-isothiocyanato-3-methylsulfanyl-benzene, acmc-20anif, 3-methylthiobenzenisothiocyanate, 3-methylthio phenylisothiocyanate, 3-methylthio-phenyl isothiocyanate, 3-methylthiophenyl isothiocyanate, 1-isothiocyanato-3-methylthio benzene
CSC1=CC=CC(=C1)N=C=S
181.271
181.28

Chemical Identifiers

51333-80-3
181.271
IHKAGFVCXPFGRP-UHFFFAOYSA-N
142841
CSC1=CC=CC(=C1)N=C=S
C8H7NS2
MFCD00041079
3-methylthio phenyl isothiocyanate, 1-isothiocyanato-3-methylsulfanyl benzene, 3-isothiocyanatophenyl methyl sulfane, 1-isothiocyanato-3-methylsulfanyl-benzene, acmc-20anif, 3-methylthiobenzenisothiocyanate, 3-methylthio phenylisothiocyanate, 3-methylthio-phenyl isothiocyanate, 3-methylthiophenyl isothiocyanate, 1-isothiocyanato-3-methylthio benzene
1-isothiocyanato-3-methylsulfanylbenzene
Safety and Handling

Safety and Handling

GHS H Statement
H331-H314-H318-H302-H312
Toxic if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Harmful in contact with skin.

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE LIQUIDS, TOXIC, N.O.S.

Documents