Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-(Methylthio)hexanol, 99%

Chemical Identifiers

Chemical Name or Material 3-(Methylthio)hexanol
CAS 51755-66-9
Molecular Formula C7H16OS
Linear Formula CH3CH2CH2CH(SCH3)CH2CH2OH
Formula Weight 148.26
MDL Number MFCD00010674
UN Number UN3334
Synonym 3-(Methylmercapto)hexanol
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2178606

 
alfa aesar™
B2178606
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAB2178614

 
alfa aesar™
B2178614
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAB2178622

 
alfa aesar™
B2178622
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 148.26
Chemical Name or Material 3-(Methylthio)hexanol
Assay 99%
Molecular Formula C7H16OS
Linear Formula CH3CH2CH2CH(SCH3)CH2CH2OH
MDL Number MFCD00010674
CAS 51755-66-9
UN Number UN3334
Synonym 3-(Methylmercapto)hexanol
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: AVIATION REGULATED LIQUID, N.O.S.
Flash Point 107°C (226°F)
Boiling Point 61° to 62°C (10mmHg)
Quantity 5g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.