missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
3-(Methylthio)butyraldehyde 95.0+%, TCI America™
Manufacturer: Tci America M193625G
Specifications
3-(Methylthio)butyraldehyde | |
Yellow | |
16630-52-7 | |
MFCD00036622 | |
NCBDFIPMWRKPDU-UHFFFAOYSA-N | |
3-methylsulfanylbutanal | |
61845 | |
Clear Liquid at 20°C |
61°C | |
25g | |
C5H10OS | |
3-methylsulfanyl butanal, 3-methylthio butanal, 3-methylthio butyraldehyde, 3-methylthiobutyraldehyde, beta-methylthio butyraldehyde, butanal, 3-methylthio, butanal,3-methylthio, butyraldehyde, 3-methylthio, fema no. 3374, potato butyraldehyde | |
CC(CC=O)SC | |
118.194 | |
118.19 | |
≥95.0% (GC) |
Chemical Identifiers
16630-52-7 | |
118.194 | |
NCBDFIPMWRKPDU-UHFFFAOYSA-N | |
61845 | |
CC(CC=O)SC |
C5H10OS | |
MFCD00036622 | |
3-methylsulfanyl butanal, 3-methylthio butanal, 3-methylthio butyraldehyde, 3-methylthiobutyraldehyde, beta-methylthio butyraldehyde, butanal, 3-methylthio, butanal,3-methylthio, butyraldehyde, 3-methylthio, fema no. 3374, potato butyraldehyde | |
3-methylsulfanylbutanal |
Safety and Handling
TSCA : Yes
Recommended Storage : Refrigerator