Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-(Methylthio)benzeneboronic acid, 97%

$59.82 - $608.73

Chemical Identifiers

Chemical Name or Material 3-(Methylthio)benzeneboronic acid
CAS 128312-11-8
Molecular Formula C7H9BO2S
Formula Weight 168.02
MDL Number MFCD01319105
Beilstein 7020728
Synonym 3-(Methylthio)phenylboronic acid; 3-Thioanisoleboronic acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL2025003

  SDS
alfa aesar™
L2025003
1g Each for $155.20

AAL2025006

  SDS
alfa aesar™
L2025006
5g Each for $608.73

AAL20250MD

  SDS
alfa aesar™
L20250MD
250mg Each for $59.82
Description & Specifications

Specifications

Formula Weight 168.02
Molecular Formula C7H9BO2S
Beilstein 7020728
CAS 128312-11-8
Health Hazard 1 Warning
Synonym 3-(Methylthio)phenylboronic acid; 3-Thioanisoleboronic acid
Chemical Name or Material 3-(Methylthio)benzeneboronic acid
MDL Number MFCD01319105
Assay 97%
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Melting Point 158° to 161°C
Quantity 1g
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.