Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-(Methylsulfonyl)benzoic acid, 98%

$86.66 - $369.43

Chemical Identifiers

Chemical Name or Material 3-(Methylsulfonyl)benzoic acid
CAS 5345-27-7
Molecular Formula C8H8O4S
Formula Weight 200.21
MDL Number MFCD00216488
Synonym 3-(Methanesulfonyl)benzoic acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5023103

 
alfa aesar™
H5023103
1g Each for $86.66

AAH5023106

 
alfa aesar™
H5023106
5g Each for $369.43
Description & Specifications

Specifications

Formula Weight 200.21
Chemical Name or Material 3-(Methylsulfonyl)benzoic acid
Molecular Formula C8H8O4S
Health Hazard 1 Warning
Melting Point 230° to 238°C
Assay 98%
MDL Number MFCD00216488
CAS 5345-27-7
Synonym 3-(Methanesulfonyl)benzoic acid
Quantity 1g
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.