Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-(Methylsulfinyl)phenylacetic acid, 96%

$509.50 - $509.50

Chemical Identifiers

Chemical Name or Material 3-(Methylsulfinyl)phenylacetic acid
CAS 1027730-68-2
Molecular Formula C9H10O3S
Formula Weight 198.24
MDL Number MFCD09953474
Synonym 3-(Methanesulfinyl)phenylacetic acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2692106

 
alfa aesar™
H2692106
5g Each for $509.50
Description & Specifications

Specifications

Formula Weight 198.24
Chemical Name or Material 3-(Methylsulfinyl)phenylacetic acid
Assay 96%
Molecular Formula C9H10O3S
MDL Number MFCD09953474
CAS 1027730-68-2
Synonym 3-(Methanesulfinyl)phenylacetic acid
Health Hazard 1 Warning
Melting Point 110° to 112°C
Quantity 5g
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.