Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Methylstyrene, 98%, stab. with 0.1% 4-tert-butylcatechol

Chemical Identifiers

Chemical Name or Material 3-Methylstyrene
CAS 100-80-1
Name Note stabilized with 0.1% 4-tert-butylcatechol
Molecular Formula C9H10
Formula Weight 118.18
MDL Number MFCD00008617
UN Number UN2618
Beilstein 1304618
Synonym 3-Vinyltoluene
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0807206

  SDS
alfa aesar™
L0807206
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Melting Point −82° to −81°C
Formula Weight 118.18
Chemical Name or Material 3-Methylstyrene
Assay 98%
Molecular Formula C9H10
MDL Number MFCD00008617
CAS 100-80-1
Beilstein 1304618
Synonym 3-Vinyltoluene
Health Hazard 1 Danger
Quantity 5g
Health Hazard 2 GHS H Statement
H304-H226-H315-H319-H335
May be fatal if swallowed and enters airways.
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P210-P301+P310-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
RTECS Number WL5075800
TSCA TSCA
Name Note stabilized with 0.1% 4-tert-butylcatechol
UN Number UN2618
Boiling Point 170° to 171°C
Flash Point 51°C (123°F)
Odor Pungent
Recommended Storage 2° to 8°C
DOT Information Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: VINYLTOLUENES, STABILIZED

Applications
3-Methylstyreneis used as pharmaceutical intermediates.

Solubility
Soluble in Methanol, Ether, Benzene, Acetone, Ethanol, Heptane. Soluble in water (0.09 g/L) at 20°C.

Notes
Store at 4°C. Protect from heat. Incompatible with oxidizing agents, heat, light, Strong acids.

Safety and Handling

Danger

GHS P Statement
P210-P301+P310-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H304-H226-H315-H319-H335
May be fatal if swallowed and enters airways.
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

2° to 8°C