Please login to your online account to display your discounted pricing

3-Methylsalicylic acid, 99%, ACROS Organics™


Quantity 100g
Packaging Plastic bottle

Manufacturer: thermo fisher scientific  AC173341000

 View more versions of this product

Catalog No. AC173341000

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Physical Form Fine Powder
Formula Weight 152.15
Assay ≥98.5%
Quantity 100g
Color Almost white
Melting Point 164° to 167°C
Beilstein 10, 220
Merck Index 15, 2570
CAS Max % 100.0
Linear Formula CH3C6H3(OH)CO2H
Assay Percent Range 99%
Molecular Formula C8H8O3
MDL Number MFCD00002448
Synonym 2-Hydroxy-3-methylbenzoic acid
CAS Min % 98.5
Health Hazard 1 Danger
Packaging Plastic bottle
Water 0.3% max. (K.F.)
Infrared Spectrum Authentic
Chemical Name or Material 3-Methylsalicylic acid
CAS 83-40-9
Health Hazard 3 GHS P Statement:
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Health Hazard 2 GHS H Statement:
Causes serious eye damage.
Harmful if swallowed.

Specifications

Chemical Name or Material 3-Methylsalicylic acid
CAS 83-40-9
CAS Min % 98.5
CAS Max % 100.0
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C8H8O3
Linear Formula CH3C6H3(OH)CO2H
Formula Weight 152.15
MDL Number MFCD00002448
Accessories

Danger

GHS P Statement:
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye damage.
Harmful if swallowed.

SDS & Safety Documents