Learn More
3-Methylsalicylic acid, 99%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 173340050
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
3-Methylsalicylic acid | |
98.5 | |
164.0°C to 167.0°C | |
Authentic | |
Glass Bottle | |
CH3C6H3(OH)CO2H | |
5 g | |
15, 2570 | |
Solubility in water: 1.5g/L (20°C). Other solubilities: soluble in alkali hydroxides, soluble in chloroform, alcohol and ether | |
CC1=CC=CC(C(O)=O)=C1O | |
152.15 | |
CHEBI:20141 | |
99% |
83-40-9 | |
100.0 | |
White | |
99% | |
C8H8O3 | |
MFCD00002448 | |
10, 220 | |
3-methylsalicylic acid, hydroxytoluic acid, o-cresotic acid, o-cresotinic acid, cresotic acid, o-homosalicylic acid, 2-hydroxy-m-toluic acid, 2,3-cresotinic acid, cresotinic acid, benzoic acid, 2-hydroxy-3-methyl | |
WHSXTWFYRGOBGO-UHFFFAOYSA-N | |
2-hydroxy-3-methylbenzoic acid | |
6738 | |
152.15 | |
Fine Powder |
Chemical Identifiers
83-40-9 | |
152.15 | |
WHSXTWFYRGOBGO-UHFFFAOYSA-N | |
6738 | |
2-hydroxy-3-methylbenzoic acid |
C8H8O3 | |
MFCD00002448 | |
3-methylsalicylic acid, hydroxytoluic acid, o-cresotic acid, o-cresotinic acid, cresotic acid, o-homosalicylic acid, 2-hydroxy-m-toluic acid, 2,3-cresotinic acid, cresotinic acid, benzoic acid, 2-hydroxy-3-methyl | |
CHEBI:20141 | |
CC1=CC=CC(C(O)=O)=C1O |
Safety and Handling
GHS H Statement
Causes serious eye damage.
Harmful if swallowed.
Causes skin irritation.
May cause respiratory irritation.
GHS P Statement
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of s
GHS Signal Word: Danger
EINECSNumber : 201-473-8
RUO – Research Use Only