missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-Methylquinoline, TCI America™

$209.31 - $2,024.34

Chemical Identifiers

CAS 612-58-8
Molecular Formula C10H9N
Molecular Weight (g/mol) 143.189
MDL Number MFCD00014661
InChI Key DTBDAFLSBDGPEA-UHFFFAOYSA-N
Synonym quinoline, 3-methyl, 3-methyl-1-benzazine, 3-methyl-quinoline, ccris 2896, unii-1yc9rbw2g0, beta-methylquinoline, 1yc9rbw2g0, 3-methyl quinoline, pubchem2344, acmc-209mql
PubChem CID 11926
ChEBI CHEBI:20140
IUPAC Name 3-methylquinoline
SMILES CC1=CC2=CC=CC=C2N=C1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
M078225G View Documents TCI America
M078225G
25g Each for $209.31
 
M0782500G View Documents TCI America
M0782500G
500g Each for $2,024.34
 
Specifications

Chemical Identifiers

612-58-8
143.189
DTBDAFLSBDGPEA-UHFFFAOYSA-N
11926
3-methylquinoline
C10H9N
MFCD00014661
quinoline, 3-methyl, 3-methyl-1-benzazine, 3-methyl-quinoline, ccris 2896, unii-1yc9rbw2g0, beta-methylquinoline, 1yc9rbw2g0, 3-methyl quinoline, pubchem2344, acmc-209mql
CHEBI:20140
CC1=CC2=CC=CC=C2N=C1

Specifications

255°C
25g
C10H9N
quinoline, 3-methyl, 3-methyl-1-benzazine, 3-methyl-quinoline, ccris 2896, unii-1yc9rbw2g0, beta-methylquinoline, 1yc9rbw2g0, 3-methyl quinoline, pubchem2344, acmc-209mql
CC1=CC2=CC=CC=C2N=C1
143.189
CHEBI:20140
Clear Liquid at 20°C
3-Methylquinoline
Yellow
612-58-8
MFCD00014661
DTBDAFLSBDGPEA-UHFFFAOYSA-N
3-methylquinoline
11926
143.19
≥98.0% (GC)
Safety and Handling

Safety and Handling

EINECSNumber : (5)-0796

RTECSNumber : VC0527000

TSCA : No