missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-Methylpyrrole, TCI America™

$438.48 - $1,127.52

Chemical Identifiers

CAS 616-43-3
Molecular Formula C5H7N
Molecular Weight (g/mol) 81.118
MDL Number MFCD00083419
InChI Key FEKWWZCCJDUWLY-UHFFFAOYSA-N
PubChem CID 12023
IUPAC Name 3-methyl-1H-pyrrole
SMILES CC1=CNC=C1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
M11441G View Documents TCI America
M11441G
1g Each for $438.48
 
M11445G View Documents TCI America
M11445G
5g Each for $1,127.52
 
Specifications

Chemical Identifiers

616-43-3
81.118
FEKWWZCCJDUWLY-UHFFFAOYSA-N
3-methyl-1H-pyrrole
C5H7N
MFCD00083419
12023
CC1=CNC=C1

Specifications

144°C
1g
C5H7N
1993
CC1=CNC=C1
81.118
81.12
≥98.0% (GC)
Yellow
616-43-3
MFCD00083419
FEKWWZCCJDUWLY-UHFFFAOYSA-N
3-methyl-1H-pyrrole
12023
Clear Liquid at 20°C
3-Methylpyrrole
Safety and Handling

Safety and Handling

TSCA : No