Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Methylpyridine-2-thiocarboxamide, 97%

$92.05 - $92.05

Chemical Identifiers

Chemical Name or Material 3-Methylpyridine-2-thiocarboxamide
Molecular Formula C7H8N2S
Formula Weight 152.21
MDL Number MFCD12026307
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH51794MD

  SDS
alfa aesar™
H51794MD
250mg Each for $92.05
Description & Specifications

Specifications

Formula Weight 152.21
Quantity 250mg
MDL Number MFCD12026307
Health Hazard 1 Danger
Chemical Name or Material 3-Methylpyridine-2-thiocarboxamide
Assay 97%
Health Hazard 2 GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Molecular Formula C7H8N2S
Melting Point 117° to 122°C
Health Hazard 3 GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Danger

GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.