missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-Methylpyridine-2-carboxaldehyde, 96%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals H5616806

 View more versions of this product

Catalog No. AAH5616806



Description

Description

It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs.

Solubility
Slightly soluble in water.

Notes
Incompatible with oxidizing agents. Store at room temperature.
Specifications

Specifications

3-Methylpyridine-2-carboxaldehyde
1.08
C7H7NO
MFCD02181144
3-methylpicolinaldehyde, 3-methyl-2-pyridinecarboxaldehyde, 3-methylpyridine-2-carboxaldehyde, 3-methyl-2-pyridinaldehyde, 2-formyl-3-picoline, 3-methyl-pyridine-2-carbaldehyde, 2-formyl-3-methylpyridine, pyridinecarboxaldehyde, methyl, 2-pyridinecarboxaldehyde, 3-methyl, 3-methyl-2-pyridineformaldehyde
JDYVLWWFVYNMTN-UHFFFAOYSA-N
3-methylpyridine-2-carbaldehyde
1501874
97%
55589-47-4
73°C (163°F)
1.538
5 g
Slightly soluble in water.
CC1=C(N=CC=C1)C=O
121.139
121.14

Chemical Identifiers

55589-47-4
121.139
JDYVLWWFVYNMTN-UHFFFAOYSA-N
1501874
CC1=C(N=CC=C1)C=O
C7H7NO
MFCD02181144
3-methylpicolinaldehyde, 3-methyl-2-pyridinecarboxaldehyde, 3-methylpyridine-2-carboxaldehyde, 3-methyl-2-pyridinaldehyde, 2-formyl-3-picoline, 3-methyl-pyridine-2-carbaldehyde, 2-formyl-3-methylpyridine, pyridinecarboxaldehyde, methyl, 2-pyridinecarboxaldehyde, 3-methyl, 3-methyl-2-pyridineformaldehyde
3-methylpyridine-2-carbaldehyde
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.

P210-P235-P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P370+P378q-P501c

H227-H302+H312+H332-H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only