missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing
3-Methylpyridazine, TCI America™
Chemical Identifiers
CAS |
1632-76-4 |
Molecular Formula |
C5H6N2 |
Molecular Weight (g/mol) |
94.117 |
MDL Number |
MFCD00006469 |
InChI Key |
MXDRPNGTQDRKQM-UHFFFAOYSA-N |
Synonym |
pyridazine, 3-methyl, methyldiazine, pyridazine, methyl, methylpyridazine, 3-methyl pyridazine, pubchem17696, 3-methylpyridazin, acmc-1bp1x, 3-methylpyridazine, ksc181a2j |
PubChem CID |
74208 |
IUPAC Name |
3-methylpyridazine |
SMILES |
CC1=NN=CC=C1 |
View More Specs
Products
2
Specifications
Chemical Identifiers
|
1632-76-4 |
|
94.117 |
|
MXDRPNGTQDRKQM-UHFFFAOYSA-N |
|
74208 |
|
CC1=NN=CC=C1 |
|
C5H6N2 |
|
MFCD00006469 |
|
pyridazine, 3-methyl, methyldiazine, pyridazine, methyl, methylpyridazine, 3-methyl pyridazine, pubchem17696, 3-methylpyridazin, acmc-1bp1x, 3-methylpyridazine, ksc181a2j |
|
3-methylpyridazine |
Specifications
|
215°C |
|
5mL |
|
C5H6N2 |
|
pyridazine, 3-methyl, methyldiazine, pyridazine, methyl, methylpyridazine, 3-methyl pyridazine, pubchem17696, 3-methylpyridazin, acmc-1bp1x, 3-methylpyridazine, ksc181a2j |
|
CC1=NN=CC=C1 |
|
94.117 |
|
94.12 |
|
≥97.0% (GC) |
|
Undesignated |
|
1632-76-4 |
|
MFCD00006469 |
|
MXDRPNGTQDRKQM-UHFFFAOYSA-N |
|
3-methylpyridazine |
|
74208 |
|
Clear Liquid at 20°C |
|
3-Methylpyridazine |
Safety and Handling