missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
3-Methylpyridazine 97.0+%, TCI America™
Supplier: TCI America M08725ML
Specifications
3-Methylpyridazine | |
215°C | |
MFCD00006469 | |
pyridazine, 3-methyl, methyldiazine, pyridazine, methyl, methylpyridazine, 3-methyl pyridazine, pubchem17696, 3-methylpyridazin, acmc-1bp1x, 3-methylpyridazine, ksc181a2j | |
CC1=NN=CC=C1 | |
94.117 | |
94.12 | |
Liquid |
1632-76-4 | |
C5H6N2 | |
5 mL | |
MXDRPNGTQDRKQM-UHFFFAOYSA-N | |
3-methylpyridazine | |
74208 | |
≥97.0% (GC) |
Chemical Identifiers
1632-76-4 | |
94.117 | |
MXDRPNGTQDRKQM-UHFFFAOYSA-N | |
74208 | |
CC1=NN=CC=C1 |
C5H6N2 | |
MFCD00006469 | |
pyridazine, 3-methyl, methyldiazine, pyridazine, methyl, methylpyridazine, 3-methyl pyridazine, pubchem17696, 3-methylpyridazin, acmc-1bp1x, 3-methylpyridazine, ksc181a2j | |
3-methylpyridazine |
Safety and Handling
TSCA : No