missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-Methylpyridazine 99%, ACROS Organics™

$157.62 - $157.62

Chemical Identifiers

CAS 1632-76-4
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 99%
Molecular Formula C5H6N2
Formula Weight 94.12
MDL Number MFCD00006469
Chemical Name or Material 3-Methylpyridazine, 99%
Assay ≥98.5%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
AC148080100 SDS Thermo Fisher Scientific
148080100
Glass bottle 10g Each for $157.62
AC148080050 SDS Thermo Fisher Scientific
148080050
Glass bottle 5g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

1632-76-4
100.0
C5H6N2
MFCD00006469
≥98.5%
98.5
99%
94.12
3-Methylpyridazine, 99%

Specifications

10g
98.5
1.0310g/mL
Glass bottle
Liquid
C5H6N2
MFCD00006469
Clear yellow to brown
≥98.5%
1632-76-4
100.0
Authentic
1.5140 to 1.5160 (20°C, 589nm)
99%
94.12
23, 92
3-Methylpyridazine, 99%

Safety and Handling

GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

EINECSNumber : 216-640-0

TSCA : TSCA