Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Methylpyridazine, 99%

$94.31 - $1,282.26

Chemical Identifiers

Chemical Name or Material 3-Methylpyridazine
CAS 1632-76-4
Molecular Formula C5H6N2
Formula Weight 94.12
MDL Number MFCD00006469
Beilstein 105691
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1409706

 
alfa aesar™
A1409706
5g Each for $94.31

AAA1409714

 
alfa aesar™
A1409714
25g Each for $349.57

AAA1409722

 
alfa aesar™
A1409722
100g Each for $1,282.26
Description & Specifications

Specifications

Formula Weight 94.12
Chemical Name or Material 3-Methylpyridazine
Assay 99%
Molecular Formula C5H6N2
MDL Number MFCD00006469
CAS 1632-76-4
Beilstein 105691
Boiling Point 213° to 214°C
Flash Point 87°C (188°F)
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
Health Hazard 3 GHS P Statement
P210-P261-P280-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Quantity 5g
Safety and Handling

Warning

GHS P Statement
P210-P261-P280-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.