Please login to your online account to display your discounted pricing

3-Methylpicolinic acid, 98%, ACROS Organics™


Quantity 5g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC335960050

 View more versions of this product

Catalog No. AC335960050

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 137.14
Physical Form Crystalline Powder
Assay ≥96.0%
Quantity 5g
Color White to tan
Melting Point 118° to 123°C
CAS 774864
CAS Max % 100.0
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
Harmful if swallowed.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Assay Percent Range 98%
Molecular Formula C7H7NO2
MDL Number MFCD00191553
Chemical Name or Material 3-Methylpicolinic acid
CAS Min % 97.5
Packaging Glass bottle
Infrared Spectrum Authentic

Specifications

Chemical Name or Material 3-Methylpicolinic acid
CAS 774864
CAS Min % 97.5
CAS Max % 100.0
Assay ≥96.0%
Assay Percent Range 98%
Molecular Formula C7H7NO2
Formula Weight 137.14
MDL Number MFCD00191553
Accessories

Warning

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
Harmful if swallowed.

SDS & Safety Documents