Please login to your online account to display your discounted pricing

(3-Methylphenyl)methanethiol, 95%, Maybridge

Chemical Identifiers

Chemical Name or Material (3-Methylphenyl)methanethiol
CAS 25697-56-7
CAS Min % 95.0
Assay 95%
Assay Percent Range 95%
Molecular Formula C8H10S
Linear Formula CH3C6H4CH2SH
Formula Weight 138.23
MDL Number MFCD00022075
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

TL00469EA

  SDS
fisher scientific company
TL00469EA
Amber glass bottle 10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 138.23
Quantity 10g
Assay 95%
CAS Min % 95.0
Packaging Amber glass bottle
Linear Formula CH3C6H4CH2SH
Health Hazard 1 GHS Signal Word: Warning
CAS 25697-56-7
Molecular Formula C8H10S
MDL Number MFCD00022075
Assay Percent Range 95%
Health Hazard 2 GHS H Statement:
Harmful in contact with skin.
Harmful if inhaled.
Harmful if swallowed.
Chemical Name or Material (3-Methylphenyl)methanethiol
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.