Please login to your online account to display your discounted pricing

(3-Methylphenyl)methanethiol, 95%, Maybridge


Quantity 10g
Packaging Amber glass bottle

Manufacturer: fisher scientific company  TL00469EA

 View more versions of this product

Catalog No. TL00469EA

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 138.23
Quantity 10g
Assay 95%
CAS Min % 95.0
Linear Formula CH3C6H4CH2SH
Packaging Amber glass bottle
Molecular Formula C8H10S
CAS 25697-56-7
Health Hazard 1 GHS Signal Word: Warning
Assay Percent Range 95%
MDL Number MFCD00022075
Health Hazard 2 GHS H Statement:
Harmful in contact with skin.
Harmful if inhaled.
Harmful if swallowed.
Chemical Name or Material (3-Methylphenyl)methanethiol
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material (3-Methylphenyl)methanethiol
CAS 25697-56-7
CAS Min % 95.0
Assay 95%
Assay Percent Range 95%
Molecular Formula C8H10S
Linear Formula CH3C6H4CH2SH
Formula Weight 138.23
MDL Number MFCD00022075
Accessories

GHS Signal Word: Warning

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful in contact with skin.
Harmful if inhaled.
Harmful if swallowed.

SDS & Safety Documents