missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

(3-Methylphenyl)(2,4,6-trimethylphenyl)iodonium Trifluoromethanesulfonate, TCI America™

$236.81 - $710.44

Chemical Identifiers

CAS 197245-87-7
Molecular Formula C17H18F3IO3S
Molecular Weight (g/mol) 486.288
MDL Number MFCD20264883
InChI Key JFTSJYFKINVLMA-UHFFFAOYSA-M
Synonym Mesityl(m-tolyl)iodonium Triflate
PubChem CID 10719708
IUPAC Name (3-methylphenyl)-(2,4,6-trimethylphenyl)iodanium;trifluoromethanesulfonate
SMILES CC1=CC(=CC=C1)[I+]C2=C(C=C(C=C2C)C)C.C(F)(F)(F)S(=O)(=O)[O-]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
M29085G View Documents TCI America
M29085G
5g Each for $710.44
 
M29081G View Documents TCI America
M29081G
1g Each for $236.81
 
Specifications

Chemical Identifiers

197245-87-7
486.288
JFTSJYFKINVLMA-UHFFFAOYSA-M
10719708
CC1=CC(=CC=C1)[I+]C2=C(C=C(C=C2C)C)C.C(F)(F)(F)S(=O)(=O)[O-]
C17H18F3IO3S
MFCD20264883
Mesityl(m-tolyl)iodonium Triflate
(3-methylphenyl)-(2,4,6-trimethylphenyl)iodanium;trifluoromethanesulfonate

Specifications

Undesignated
5g
C17H18F3IO3S
Mesityl(m-tolyl)iodonium Triflate
CC1=CC(=CC=C1)[I+]C2=C(C=C(C=C2C)C)C.C(F)(F)(F)S(=O)(=O)[O-]
486.288
486.29
≥98.0% (HPLC,T)
168°C
197245-87-7
MFCD20264883
JFTSJYFKINVLMA-UHFFFAOYSA-M
(3-methylphenyl)-(2,4,6-trimethylphenyl)iodanium;trifluoromethanesulfonate
10719708
Crystal-Powder at 20°C
(3-Methylphenyl)(2,4,6-trimethylphenyl)iodonium Trifluoromethanesulfonate
Safety and Handling

Safety and Handling

TSCA : No