Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Methylphenoxyacetic acid, 98%

$123.55 - $1,654.42

Chemical Identifiers

Chemical Name or Material 3-Methylphenoxyacetic acid
CAS 1643-15-8
Molecular Formula C9H10O3
Formula Weight 166.18
MDL Number MFCD00014358
Beilstein 2047572
Synonym m-Cresoxyacetic acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2486906

  SDS
alfa aesar™
B2486906
5g Each for $123.55

AAB2486914

  SDS
alfa aesar™
B2486914
25g Each for $474.64

AAB2486922

  SDS
alfa aesar™
B2486922
100g Each for $1,654.42
Description & Specifications

Specifications

Formula Weight 166.18
Health Hazard 1 Warning
Chemical Name or Material 3-Methylphenoxyacetic acid
Assay 98%
Beilstein 2047572
Quantity 5g
CAS 1643-15-8
Synonym m-Cresoxyacetic acid
Melting Point 102° to 105°C
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Molecular Formula C9H10O3
RTECS Number AI9316500
MDL Number MFCD00014358

Applications
3-Methylphenoxyacetic acid is used as pharmaceutical intermediates.

Solubility
Soluble in water. 4810 mg/L at 25°C

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible materials are oxidizing agents.

Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.