Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Methyloxindole, 97%

$59.07 - $59.07

Chemical Identifiers

Chemical Name or Material 3-Methyloxindole
CAS 1504-06-9
Molecular Formula C9H9NO
Formula Weight 147.18
MDL Number MFCD04037370
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2766206

  SDS
alfa aesar™
H2766206
5g Each for $59.07
Description & Specifications

Specifications

Formula Weight 147.18
Chemical Name or Material 3-Methyloxindole
Assay 97%
Molecular Formula C9H9NO
MDL Number MFCD04037370
CAS 1504-06-9
Melting Point 121° to 124°C
Quantity 5g

Applications
A versatile reactant. As 3-Methylindole metabolite. Reactant for enantioselective α-amination reactions, Reactant for aldol reaction with glyoxal derivatives, Reactant for amine thiourea catalyzed conjugate addition to α,β-unsaturated aldehydes, Reactant for O-acetylation reactions, Reactant for preparation of a disubstituted oxoindole by using rhodium-catalyzed cyclopropanation/ring-opening reactions.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.