missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Methyloxetane-3-carboxylic acid, 96%

$73.11 - $223.61

Chemical Identifiers

CAS 28562-68-7
Molecular Formula C5H8O3
Molecular Weight (g/mol) 116.116
MDL Number MFCD09842435
InChI Key PMPZEEUNGQSAIQ-UHFFFAOYSA-N
Synonym 3-methyloxetane-3-carboxylicacid, 3-methyl-3-oxetanecarboxylic acid, 3-oxetanecarboxylic acid, 3-methyl, 3-methyl-oxetane-3-carboxylic acid, pubchem14214, ksc199o1l, 3-methyl oxetane-3-carboxylic acid, 3-methyl-3-oxetane carboxylic acid, 3-oxetanecarboxylicacid, 3-methyl, 3-methyloxetane-3-carboxylic acid
PubChem CID 14028997
IUPAC Name 3-methyloxetane-3-carboxylic acid
SMILES CC1(COC1)C(=O)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH66283MD View Documents Alfa Aesar™
H66283MD
250mg Each for $73.11
 
AAH6628303 View Documents Alfa Aesar™
H6628303
1g Each for $223.61
 
Specifications

Chemical Identifiers

28562-68-7
116.116
PMPZEEUNGQSAIQ-UHFFFAOYSA-N
14028997
CC1(COC1)C(=O)O
C5H8O3
MFCD09842435
3-methyloxetane-3-carboxylicacid, 3-methyl-3-oxetanecarboxylic acid, 3-oxetanecarboxylic acid, 3-methyl, 3-methyl-oxetane-3-carboxylic acid, pubchem14214, ksc199o1l, 3-methyl oxetane-3-carboxylic acid, 3-methyl-3-oxetane carboxylic acid, 3-oxetanecarboxylicacid, 3-methyl, 3-methyloxetane-3-carboxylic acid
3-methyloxetane-3-carboxylic acid

Specifications

Moisture Sensitive
58°C to 63°C
28562-68-7
MFCD09842435
PMPZEEUNGQSAIQ-UHFFFAOYSA-N
3-methyloxetane-3-carboxylic acid
14028997
Powder
3-Methyloxetane-3-carboxylic acid
White to Yellow
250mg
C5H8O3
3-methyloxetane-3-carboxylicacid, 3-methyl-3-oxetanecarboxylic acid, 3-oxetanecarboxylic acid, 3-methyl, 3-methyl-oxetane-3-carboxylic acid, pubchem14214, ksc199o1l, 3-methyl oxetane-3-carboxylic acid, 3-methyl-3-oxetane carboxylic acid, 3-oxetanecarboxylicacid, 3-methyl, 3-methyloxetane-3-carboxylic acid
CC1(COC1)C(=O)O
116.116
116.12
96%
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

storageNote1 : -30° to -10°C