missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-Methylisoxazole-5-carbaldehyde, 95%, Maybridge

Chemical Identifiers

CAS 70753-36-5
CAS Min % 94.5
CAS Max % 100.0
Assay Percent Range 95%
Molecular Formula C5H5NO2
Formula Weight 111.1
MDL Number MFCD09751673
Chemical Name or Material 3-Methylisoxazole-5-carbaldehyde
Assay 95%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
CC27004CB SDS Fisher Scientific Company
CC27004CB
Amber Glass Bottle 250mg N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
CC27004DA SDS Fisher Scientific Company
CC27004DA
Amber Glass Bottle 1g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

70753-36-5
100.0
C5H5NO2
MFCD09751673
95%
94.5
95%
111.1
3-Methylisoxazole-5-carbaldehyde

Specifications

111.1
100.0
70753-36-5
MFCD09751673
95%
95%
250mg
C5H5NO2
94.5
Amber Glass Bottle
3-Methylisoxazole-5-carbaldehyde

Safety and Handling

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Harmful if swallowed.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning