Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Methylisoxazole-4-carboxylic acid, 98+%

$83.01 - $248.62

Chemical Identifiers

Chemical Name or Material 3-Methylisoxazole-4-carboxylic acid
CAS 17153-20-7
Molecular Formula C5H5NO3
Formula Weight 127.1
MDL Number MFCD01318161
Beilstein 114157
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1630703

  SDS
alfa aesar™
L1630703
1g Each for $248.62

AAL16307MD

  SDS
alfa aesar™
L16307MD
250mg Each for $83.01
Description & Specifications

Specifications

Formula Weight 127.1
Chemical Name or Material 3-Methylisoxazole-4-carboxylic acid
Assay ≥98%
Molecular Formula C5H5NO3
Beilstein 114157
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
MDL Number MFCD01318161
CAS 17153-20-7
Melting Point 185° to 187°C
Quantity 1g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.