missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-Methylindole, TCI America™

Manufacturer:  TCI America M034725G

 View more versions of this product

Catalog No. M034725G



Specifications

Specifications

3-Methylindole
White-Yellow
25g
C9H9N
3-methylindole, skatole, scatole, skatol, 1h-indole, 3-methyl, beta-methylindole, indole, 3-methyl, 3-mi, 3-methyl-4,5-benzopyrrole, 3-methyl indole
CC1=CNC2=CC=CC=C12
131.178
CHEBI:9171
Crystal-Powder at 20°C
141°C
96°C
83-34-1
MFCD00005627
ZFRKQXVRDFCRJG-UHFFFAOYSA-N
3-methyl-1H-indole
6736
131.18
≥98.0% (GC)

Chemical Identifiers

83-34-1
131.178
ZFRKQXVRDFCRJG-UHFFFAOYSA-N
6736
3-methyl-1H-indole
C9H9N
MFCD00005627
3-methylindole, skatole, scatole, skatol, 1h-indole, 3-methyl, beta-methylindole, indole, 3-methyl, 3-mi, 3-methyl-4,5-benzopyrrole, 3-methyl indole
CHEBI:9171
CC1=CNC2=CC=CC=C12
Safety and Handling

Safety and Handling

RTECSNumber : NM0350000

TSCA : Yes

Documents