missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Methylheptane, 97%

Manufacturer:  Alfa Aesar™ L1200614

 View more versions of this product

Catalog No. AAL1200614



Specifications

Specifications

3-Methylheptane
118°C to 120°C
25g
C8H18
UN1262
heptane, 3-methyl, 2-ethylhexane, 3-methyl-heptane, 2-butylbutane, 3-methyl-heptan, heptane, 3-methyl-, r, acmc-20mdw0, 3-methyl-s-heptane, 3-methylheptane, acmc-1axq2
CCCCC(C)CC
114.232
114.23
7°C (44°F)
-121°C
589-81-1
MFCD00027244
1730777
LAIUFBWHERIJIH-UHFFFAOYSA-N
3-methylheptane
11519
97%

Chemical Identifiers

589-81-1
114.232
LAIUFBWHERIJIH-UHFFFAOYSA-N
11519
CCCCC(C)CC
C8H18
MFCD00027244
heptane, 3-methyl, 2-ethylhexane, 3-methyl-heptane, 2-butylbutane, 3-methyl-heptan, heptane, 3-methyl-, r, acmc-20mdw0, 3-methyl-s-heptane, 3-methylheptane, acmc-1axq2
3-methylheptane
Safety and Handling

Safety and Handling

GHS H Statement
H225-H304-H315-H336
Highly flammable liquid and vapor.
May be fatal if swallowed and enters airways.
Causes skin irritation.
May cause drowsiness or dizziness.

GHS P Statement
P210-P261-P301+P310-P303+P361+P353-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: OCTANES

TSCA : TSCA

Documents