Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Methylglutaric acid, 95%

$57.57 - $57.57

Chemical Identifiers

Chemical Name or Material 3-Methylglutaric acid
CAS 626-51-7
Molecular Formula C6H10O4
Formula Weight 146.14
MDL Number MFCD00002727
Beilstein 1759502
Synonym 3-Methylpropane-1,3-dicarboxylic acid; 3-Methylpentanedioic acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2473706

  SDS
alfa aesar™
B2473706
5g Each for $57.57
Description & Specifications

Specifications

Formula Weight 146.14
Quantity 5g
Molecular Formula C6H10O4
Beilstein 1759502
Chemical Name or Material 3-Methylglutaric acid
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Melting Point 78° to 83°C
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Assay 95%
CAS 626-51-7
Synonym 3-Methylpropane-1,3-dicarboxylic acid; 3-Methylpentanedioic acid
Health Hazard 1 Warning
MDL Number MFCD00002727

Applications
3-Methylglutaric acid is a hypolipidemic agent and also used as an chemical, organic intermediate, pharmaceutical intermediate.

Solubility
Soluble in water, and ethanol.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.

Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.