missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-Methylfuran (stabilized with HQ) 98.0+%, TCI America™

Manufacturer:  TCI America M09395G

 View more versions of this product

Catalog No. M09395G



Specifications

Specifications

3-Methylfuran (stabilized with HQ)
Yellow
930-27-8
MFCD00060134
furan, 3-methyl, 3-methylfurane, 3-methyl furan, 3-methyl-furan, 3-methyl-fura, 3-methylfuran, pubchem19993, 3-methylfuran-methyl-d3, acmc-20an04, 3-methylfuran 1gr
CC1=COC=C1
82.102
82.10
≥98.0% (GC)
66°C
5g
C5H6O
1993
KJRRQXYWFQKJIP-UHFFFAOYSA-N
3-methylfuran
13587
Clear Liquid at 20°C

Chemical Identifiers

930-27-8
82.102
KJRRQXYWFQKJIP-UHFFFAOYSA-N
13587
CC1=COC=C1
C5H6O
MFCD00060134
furan, 3-methyl, 3-methylfurane, 3-methyl furan, 3-methyl-furan, 3-methyl-fura, 3-methylfuran, pubchem19993, 3-methylfuran-methyl-d3, acmc-20an04, 3-methylfuran 1gr
3-methylfuran
Safety and Handling

Safety and Handling

RTECSNumber : LU2630000

TSCA : No

Recommended Storage : Refrigerator

Documents