missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-Methylfuran (stabilized with HQ), TCI America™

$398.77 - $1,287.94

Chemical Identifiers

CAS 930-27-8
Molecular Formula C5H6O
Molecular Weight (g/mol) 82.102
MDL Number MFCD00060134
InChI Key KJRRQXYWFQKJIP-UHFFFAOYSA-N
Synonym furan, 3-methyl, 3-methylfurane, 3-methyl furan, 3-methyl-furan, 3-methyl-fura, 3-methylfuran, pubchem19993, 3-methylfuran-methyl-d3, acmc-20an04, 3-methylfuran 1gr
PubChem CID 13587
IUPAC Name 3-methylfuran
SMILES CC1=COC=C1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
M09391G View Documents TCI America
M09391G
1g Each for $398.77
 
M09395G View Documents TCI America
M09395G
5g Each for $1,287.94
 
Specifications

Chemical Identifiers

930-27-8
82.102
KJRRQXYWFQKJIP-UHFFFAOYSA-N
13587
CC1=COC=C1
C5H6O
MFCD00060134
furan, 3-methyl, 3-methylfurane, 3-methyl furan, 3-methyl-furan, 3-methyl-fura, 3-methylfuran, pubchem19993, 3-methylfuran-methyl-d3, acmc-20an04, 3-methylfuran 1gr
3-methylfuran

Specifications

66°C
1g
C5H6O
1993
KJRRQXYWFQKJIP-UHFFFAOYSA-N
3-methylfuran
13587
Clear Liquid at 20°C
3-Methylfuran (stabilized with HQ)
Yellow
930-27-8
MFCD00060134
furan, 3-methyl, 3-methylfurane, 3-methyl furan, 3-methyl-furan, 3-methyl-fura, 3-methylfuran, pubchem19993, 3-methylfuran-methyl-d3, acmc-20an04, 3-methylfuran 1gr
CC1=COC=C1
82.102
82.10
≥98.0% (GC)
Safety and Handling

Safety and Handling

RTECSNumber : LU2630000

TSCA : No

Recommended Storage : Refrigerator