Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Methylformanilide, 97%

$54.39 - $714.20

Chemical Identifiers

Chemical Name or Material 3-Methylformanilide
CAS 3085-53-8
Molecular Formula C8H9NO
Formula Weight 135.17
MDL Number MFCD00014123
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2406106

  SDS
alfa aesar™
B2406106
5g Each for $54.39

AAB2406114

  SDS
alfa aesar™
B2406114
25g Each for $213.96

AAB2406122

  SDS
alfa aesar™
B2406122
100g Each for $714.20
Description & Specifications

Specifications

Formula Weight 135.17
Chemical Name or Material 3-Methylformanilide
Assay 97%
Molecular Formula C8H9NO
MDL Number MFCD00014123
CAS 3085-53-8
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H301
Toxic if swallowed.
Health Hazard 3 GHS P Statement
P264-P270-P301+P310-P321-P405-P501a
Wash thoroughly after handling.
Do not eat, drink or smoke when using this product.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Specific treatment (see label).
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Boiling Point 165° to 167°C (10mmHg)
Quantity 5g
Safety and Handling

Danger

GHS P Statement
P264-P270-P301+P310-P321-P405-P501a
Wash thoroughly after handling.
Do not eat, drink or smoke when using this product.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Specific treatment (see label).
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301
Toxic if swallowed.