Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Methylflavone-8-carboxylic acid, 98+%

$213.96 - $213.96

Chemical Identifiers

Chemical Name or Material 3-Methylflavone-8-carboxylic acid
CAS 3468-01-7
Molecular Formula C17H12O4
Formula Weight 280.28
MDL Number MFCD00040998
Beilstein 1433509
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1132014

  SDS
alfa aesar™
L1132014
25g Each for $213.96
Description & Specifications

Specifications

Formula Weight 280.28
CAS 3468-01-7
MDL Number MFCD00040998
Chemical Name or Material 3-Methylflavone-8-carboxylic acid
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Assay ≥98%
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Molecular Formula C17H12O4
Quantity 25g
Melting Point 234° to 236°C
Beilstein 1433509
Health Hazard 1 Warning
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.