missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Methylflavone-8-carboxylic acid, 98+%

Manufacturer:  Alfa Aesar™ L1132006

 View more versions of this product

Catalog No. AAL1132006



Specifications

Specifications

3-Methylflavone-8-carboxylic acid
5g
C10H10O
1433509
PJRHFTYXYCVOSJ-UHFFFAOYSA-N
cyclopropyl(phenyl)methanone
77029
≥98%
234°C to 236°C
3481-02-5
MFCD00040998
cyclopropyl phenyl ketone, cyclopropyl phenyl methanone, phenyl cyclopropyl ketone, benzoylcyclopropane, methanone, cyclopropylphenyl, cyclopropylphenylketone, ketone, cyclopropyl phenyl, cyclopropyl-phenylmethanone, acmc-2097dl, cyclopropyl-phenyl-methanone
C1CC1C(=O)C2=CC=CC=C2
146.19
280.28

Chemical Identifiers

3481-02-5
146.19
PJRHFTYXYCVOSJ-UHFFFAOYSA-N
77029
C1CC1C(=O)C2=CC=CC=C2
C10H10O
MFCD00040998
cyclopropyl phenyl ketone, cyclopropyl phenyl methanone, phenyl cyclopropyl ketone, benzoylcyclopropane, methanone, cyclopropylphenyl, cyclopropylphenylketone, ketone, cyclopropyl phenyl, cyclopropyl-phenylmethanone, acmc-2097dl, cyclopropyl-phenyl-methanone
cyclopropyl(phenyl)methanone
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

Documents