missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Methylflavone-8-carboxylic acid, 98+%

Manufacturer:  Alfa Aesar™ L1132014

 View more versions of this product

Catalog No. AAL1132014



Specifications

Specifications

3-Methylflavone-8-carboxylic acid
25g
C17H12O4
1433509
KMMBBZOSQNLLMN-UHFFFAOYSA-N
3-methyl-4-oxo-2-phenylchromene-8-carboxylic acid
77016
280.28
234°C to 236°C
3468-01-7
MFCD00040998
3-methylflavone-8-carboxylic acid, 3-methyl-4-oxo-2-phenyl-4h-chromene-8-carboxylic acid, 8-carboxy-3-methylflavone, 4h-1-benzopyran-8-carboxylicacid, 3-methyl-4-oxo-2-phenyl, unii-o8rcm70c48, 4h-1-benzopyran-8-carboxylic acid, 3-methyl-4-oxo-2-phenyl, 3-methyl-4-oxo-2-phenyl-4h-1-benzopyran-8-carboxylic acid, enamine_005545, 3-methylflavone-8-carboxylicacid, acmc-20a6bq
CC1=C(OC2=C(C1=O)C=CC=C2C(=O)O)C3=CC=CC=C3
280.28
CHEBI:61237
≥98%

Chemical Identifiers

3468-01-7
280.28
KMMBBZOSQNLLMN-UHFFFAOYSA-N
77016
3-methyl-4-oxo-2-phenylchromene-8-carboxylic acid
C17H12O4
MFCD00040998
3-methylflavone-8-carboxylic acid, 3-methyl-4-oxo-2-phenyl-4h-chromene-8-carboxylic acid, 8-carboxy-3-methylflavone, 4h-1-benzopyran-8-carboxylicacid, 3-methyl-4-oxo-2-phenyl, unii-o8rcm70c48, 4h-1-benzopyran-8-carboxylic acid, 3-methyl-4-oxo-2-phenyl, 3-methyl-4-oxo-2-phenyl-4h-1-benzopyran-8-carboxylic acid, enamine_005545, 3-methylflavone-8-carboxylicacid, acmc-20a6bq
CHEBI:61237
CC1=C(OC2=C(C1=O)C=CC=C2C(=O)O)C3=CC=CC=C3
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

Documents