Please login to your online account to display your discounted pricing

3-Methylenecyclopropane-trans-1,2-dicarboxylic acid, 98%, ACROS Organics™


Quantity 1g
Packaging Glass bottle

Manufacturer: fisher scientific acros  205290010

 View more versions of this product

Catalog No. AC205290010

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Infrared Spectrum Authentic
Packaging Glass bottle
Melting Point 197° to 201°C
Color Off-white to beige
Quantity 1g
Chemical Name or Material 3-Methylenecyclopropane-trans-1, 2-dicarboxylic acid
Name Note 98%
CAS 499-02-5
Physical Form Off-white to beige crystalline powder
Assay 97.5% min.
Assay Percent Range 98%
Molecular Formula C6H6O4
Formula Weight 142.11g/mol
MDL Number MFCD00001312
Beilstein 09, IV, 3935
Health Hazard 2 GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.
Health Hazard 3 GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECS Number 207-874-4

Specifications

Chemical Name or Material 3-Methylenecyclopropane-trans-1, 2-dicarboxylic acid
Name Note 98%
CAS 499-02-5
Assay 97.5% min.
Assay Percent Range 98%
Molecular Formula C6H6O4
Formula Weight 142.11g/mol
MDL Number MFCD00001312
Accessories

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.

SDS & Safety Documents