Please login to your online account to display your discounted pricing

3-Methylenecyclopropane-trans-1,2-dicarboxylic acid, 98%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 3-Methylenecyclopropane-trans-1, 2-dicarboxylic acid
Name Note 98%
CAS 499-02-5
Assay 97.5% min.
Assay Percent Range 98%
Molecular Formula C6H6O4
Formula Weight 142.11g/mol
MDL Number MFCD00001312
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC205290010

  SDS
fisher scientific acros
205290010
Glass bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC205292500

  SDS
thermo fisher scientific
205292500
Glass bottle 250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Infrared Spectrum Authentic
Packaging Glass bottle
Melting Point 197° to 201°C
Color Off-white to beige
Quantity 1g
Chemical Name or Material 3-Methylenecyclopropane-trans-1, 2-dicarboxylic acid
Name Note 98%
CAS 499-02-5
Physical Form Off-white to beige crystalline powder
Assay 97.5% min.
Assay Percent Range 98%
Molecular Formula C6H6O4
Formula Weight 142.11g/mol
MDL Number MFCD00001312
Beilstein 09, IV, 3935
Health Hazard 2 GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.
Health Hazard 3 GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECS Number 207-874-4
Safety and Handling

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.