Please login to your online account to display your discounted pricing

3-Methylenecyclobutanecarbonitrile, 97%, Maybridge

Chemical Identifiers

Chemical Name or Material 3-Methylenecyclobutanecarbonitrile
CAS 15760-35-7
CAS Min % 97.0
Assay 97%
Assay Percent Range 97%
Molecular Formula C6H7N
Linear Formula CH2=C4H5CN
Formula Weight 93.13
MDL Number MFCD01085401
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

SB02040DA

  SDS
fisher scientific company
SB02040DA
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 93.13
Quantity 1g
Assay 97%
CAS Min % 97.0
MDL Number MFCD01085401
Molecular Formula C6H7N
Linear Formula CH2=C4H5CN
Health Hazard 1 Warning
Chemical Name or Material 3-Methylenecyclobutanecarbonitrile
Health Hazard 2 GHS P Statement Harmful if swallowed. Harmful if inhaled. Harmful in contact with skin.
CAS 15760-35-7
Assay Percent Range 97%
Health Hazard 3 GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. IF ON SKIN: Wash with plenty of soap and water. Call a POISON CENTER or doctor/physician if you feel unwell. IF INHALED: Remov