Please login to your online account to display your discounted pricing

3-Methylenecyclobutanecarbonitrile, 97%, Maybridge


Quantity 1g

Manufacturer: fisher scientific company  SB02040DA

 View more versions of this product

Catalog No. SB02040DA

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 93.13
Quantity 1g
Assay 97%
CAS Min % 97.0
Molecular Formula C6H7N
MDL Number MFCD01085401
Chemical Name or Material 3-Methylenecyclobutanecarbonitrile
Linear Formula CH2=C4H5CN
Health Hazard 1 Warning
CAS 15760-35-7
Health Hazard 2 GHS P Statement Harmful if swallowed. Harmful if inhaled. Harmful in contact with skin.
Assay Percent Range 97%
Health Hazard 3 GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. IF ON SKIN: Wash with plenty of soap and water. Call a POISON CENTER or doctor/physician if you feel unwell. IF INHALED: Remov

Specifications

Chemical Name or Material 3-Methylenecyclobutanecarbonitrile
CAS 15760-35-7
CAS Min % 97.0
Assay 97%
Assay Percent Range 97%
Molecular Formula C6H7N
Linear Formula CH2=C4H5CN
Formula Weight 93.13
MDL Number MFCD01085401
Accessories

Warning

GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. IF ON SKIN: Wash with plenty of soap and water. Call a POISON CENTER or doctor/physician if you feel unwell. IF INHALED: Remov

GHS P Statement Harmful if swallowed. Harmful if inhaled. Harmful in contact with skin.